EMAIL THIS PAGE TO A FRIEND
SRP6041 Sigma-Aldrich

IL-17A/F human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym: IL-17 A/F, IL17 A/F, Interleukin-17 A/F, Interleukin-17 AF

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
biological source   human
recombinant   expressed in E. coli
assay   ≥98% (SDS-PAGE)
form   lyophilized
mol wt   mol wt 30.7 kDa
packaging   pkg of 10 μg
impurities   Endotoxin, tested
NCBI accession no.   IL17A: Q16552
  IL17F: Q96PD4
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... IL17A(3605), IL17F(112744)

Description

General description

Human IL-17A/F is a 40 kDa glycoprotein which is secreted as a disulfide-linked heterodimer. IL-17A/F consists of two proteins, IL-17A and IL17F. Human IL17A is produced as a 155 a.a precursor that includes a 23 amino acids signal sequence and a 132 amino acid chain that includes an N-linked glycosylation site. Human IL17F is produced as a 153 amino acid precursor with a 20 amino acid signal sequence and a 133 amino acid region and an N-linked glycosylation site. Both proteins (IL17A & IL17F) share 50 % amino acid sequence identity. Human IL17A & IL17F show approximately 60 % homology in their amino acid sequence to mouse IL-17A and IL-17F. Interleukin-17A/F and IL17A, IL17F homodimers are manufactured by activted CD4+ T cells, called Th17. IL-23 causes Th17 lymphocytes to manufacture IL-17A/F. Interleukin-17A/F induces chemokine production and airway neutrophilia with intermediate potency between IL17A (most potent) and IL17F (least potent). IL-17A/F Human Recombinant produced in E.Coli is a heterodimeric, non-glycosylated polypeptide chain containing 1 monomeric subunit of each IL-17A & IL-17F. The active dimer contains 271 amino acids and having a total molecular mass of 30.7 kDa. The IL-17A/F Human is purified by proprietary chromatographic techniques.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute with sterile H?O to a concentration not more than 1 mg/ml; This solution can then be diluted into other aqueous buffers.

Physical form

Lyophilized with no additives

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Request a Serum Sample

Cell Culture Guide
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?