• USA Home
  • SRP6070 - Cyclophilin F human

EMAIL THIS PAGE TO A FRIEND
SRP6070 Sigma-Aldrich

Cyclophilin F human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym: CYP3, Cyp-D, PPIF, Peptidyl prolyl isomerase F

  •  NACRES NA.32

Purchase

Properties

biological source   human
recombinant   expressed in E. coli
assay   ≥95% (SDS-PAGE)
form   liquid
mol wt   21 kDa
packaging   pkg of 100 μg
NCBI accession no.   P30405
UniProt accession no.   P30405
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... PPIF(10105)

Description

General description

Cyclophilin F is a member of peptidyl-prolyl cis-trans isomerase family. They are highly-conserved cytoplasmic enzymes that accelerate protein folding. It is a part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 7.5) containing 1 mM DTT 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH PSFMCQAGDF TNHNGTGGKS IYGSRFPDEN FTLKHVGPGV LSMANAGPNT NGSQFFICTI KTDWLDGKHV VFGHVKEGMD VVKKIESFGS KSGRTSKKIV ITDCGQLS

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?