EMAIL THIS PAGE TO A FRIEND
SRP6183 Sigma-Aldrich

FGF-4 human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE)

Synonym: HBGF-4, HST-1, HST, HSTF1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, K-FGF, KFGF, Transforming protein KS3

Purchase

Properties

Related Categories Cell Culture, FGF-4 to FGF-8, Fibroblast Growth Factors (FGF), Growth Factors and Cytokines, Reagents and Supplements More...
biological source   human
recombinant   expressed in E. coli
assay   ≥97% (SDS-PAGE)
form   lyophilized
mol wt   mol wt 19 kDa
packaging   pkg of 5 mg
UniProt accession no.   P08620
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... FGF-4(2249)

Description

Reconstitution

When reconstituting the product, gently pipet and wash down the sides of the vial to ensure full recovery of the protein into solution. It is recommended to reconstitute the lyophilized product with sterile water at a concentration of 0.1 mg/mL, which can be further diluted into other aqueous solutions.

Physical form

Lyophilized from a sterile solution containing 10 mM Sodium phosphate buffer and 75 mM NaCl. .

General description

Fibroblast Growth Factor 4 (FGF-4) is a growth factor predominantly expressed during embryonic development, playing a key role in limb development. In culture, FGF-4 has been shown to be an important growth factor for fibroblasts and endothelial cells. Human FGF-4 shares high homology and cross-reactivity with the mouse protein. Recombinant human FGF-4, produced in E.coli, is a non-glycosylated protein containing 177 amino acids and having a total molecular mass of 19 kDa.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis


Cell Culture Guide
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?