EMAIL THIS PAGE TO A FRIEND
SRP6398 Sigma-Aldrich

MSR1/CD204 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym: SCARA1, SR-A, phSR1, phSR2

Purchase

Properties

Related Categories CD160 to CD340, Cell Biology, Cell Signaling and Neuroscience, Cluster of Differentiation (CD) Proteins, Cytokines and Growth Factors,
biological source   human
recombinant   expressed in HEK 293 cells
assay   ≥95% (SDS-PAGE)
form   lyophilized
mol wt   calculated mol wt 42.1 kDa
  observed mol wt 65-72 kDa (DTT-reduced. Protein migrates due to glycosylation. Lys 77 is the predicted N-terminus.)
packaging   pkg of 10 and 50 μg
impurities   <1 EU/μg endotoxin (LAL test)
conjugate   6-His tagged (C-terminus)
UniProt accession no.   P21757
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MSR1(4481)

Description

General description

Macrophage scavenger receptor 1 (MSR1) also known as CD204, SCARA1, SR-A, and is class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. Defects in MSR1 may be a cause of prostate cancer (PC) or Barrett esophagus (BE).

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?