• USA Home
  • T0563 - Anti-Transforming Growth Factor-α antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
T0563 Sigma

Anti-Transforming Growth Factor-α antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-TGF-α

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Cytokines, Chemokines, and Growth Factors for Stem Cell Biology, Antibodies against Cytokines/Growth Factors/Chemokines for Stem Cell Biology, Antibodies for Cell Biology,
species reactivity   human
application(s)   capture ELISA: suitable
  immunohistochemistry: 15 μg/mL using using cells and tissue with antigen retrieval and chromogenic detection system
  neutralization: suitable
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... TGFA(7039)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant human TGF-α.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline and 5% trehalose.

Specificity

The antibody will neutralize the biological activity of recombinant human TGF-α.

Preparation Note

Affinity isolated using human TGF-α affinity chromatography.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?