• USA Home
  • T1449 - Trizma® Buffer solution

EMAIL THIS PAGE TO A FRIEND
T1449 Sigma-Aldrich

Trizma® Buffer solution

0.3 M

Synonym: tris buffer solution

  •  eCl@ss 32129211

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions, Buffers A to Z More...
concentration   0.3 M
pH   8.5 (37 °C)
  8.8 (25 °C)
  9.4 (5 °C)
storage temp.   2-8°C

Description

Suitability

Suitable for use as a reconstitution buffer in enzymatic assays such as G-6-PDH determination. Contains chloroform at 0.2% as a preservative.

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?