EMAIL THIS PAGE TO A FRIEND
T3501 Sigma-Aldrich

o-Tolidine

practical grade

Synonym: 3,3′-Dimethylbenzidine, 4,4′-Bianisidine

  • CAS Number 119-93-7

  • Linear Formula [-C6H3(CH3)-4-NH2]2

  • Molecular Weight 212.29

  •  Beilstein/REAXYS Number 2210640

  •  EC Number 204-358-0

  •  MDL number MFCD00014773

Purchase

Properties

grade   practical grade
mp   128-131 °C (lit.)
SMILES string   Cc1cc(ccc1N)-c2ccc(N)c(C)c2
InChI   1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3
InChI key   NUIURNJTPRWVAP-UHFFFAOYSA-N

Description

Application

Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
DD1225000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?