• USA Home
  • T3771 - 2-Thiopheneacetonitrile

EMAIL THIS PAGE TO A FRIEND
T3771 SAFC

2-Thiopheneacetonitrile

~97%

Purchase

Properties

assay   ~97%
density   1.16 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 3276 6.1/PG 3
WGK Germany 
3
Flash Point(F) 
215 °F
Flash Point(C) 
102 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?