• USA Home
  • T4147 - Trehalose octaacetate

EMAIL THIS PAGE TO A FRIEND
T4147 Sigma-Aldrich

Trehalose octaacetate

~99%

  • CAS Number 25018-27-3

  • Empirical Formula (Hill Notation) C28H38O19

  • Molecular Weight 678.59

Purchase

Properties

assay   ~99%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number

Certificate of Origin

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?