EMAIL THIS PAGE TO A FRIEND
T431060 Sigma-Aldrich

DMT-dT-CPG column

pore size 1000 Å, 1 μmol, configured for PerkinElmer, configured for ABI

Purchase

Properties

Related Categories ABI 39X, Chemical Synthesis, DNA Column, Expedite, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
InChI Key   JUNZDJNCZJUDRU-UHFFFAOYSA-N
type   for DNA synthesis
product line   Proligo Reagents
assay   ≥98.0% (Nucleoside Purity, reversed phase HPLC)
pore size   1000 Å
capacity   ≥99 % (Coupling Efficiency)
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
  configured for PerkinElmer
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?