• USA Home
  • T4406 - Δ9-Tetrahydrocannabinol-d3

EMAIL THIS PAGE TO A FRIEND
T4406 Supelco

Δ9-Tetrahydrocannabinol-d3

ethanol solution

Synonym: Δ1-Tetrahydrocannabinol-d3, 6α,7,8,10α-Tetrahydro-6,6,9-trimethyl-3-(pentyl-5,5,5-d3)-6H-dibenzo[b,d]pyran-1-ol

  • CAS Number 81586-39-2

  • Empirical Formula (Hill Notation) C21H27D3O2

  • Molecular Weight 317.48

  •  MDL number MFCD00083507

Purchase

Properties

isotopic purity   98 atom % D
form   ethanol solution
  neat
drug control   USDEA Schedule I; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 2

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?