• USA Home
  • T4901 - Tresyl chloride-activated agarose

EMAIL THIS PAGE TO A FRIEND
T4901 Sigma-Aldrich

Tresyl chloride-activated agarose

lyophilized powder

Purchase

Properties

form   lyophilized powder
matrix   Cross-linked 4% beaded agarose
matrix active group   tresyl (2,2,2-trifluoroethanesulfonyl)
matrix spacer   (direct linkage to agarose (no spacer) by displacement of tresyl group)
capacity   ~8 mg/mL coupling capacity (chymotrypsin)(covalent)
storage temp.   2-8°C

Description

Quantity

Swelling ratio: 1 g swells to approx. 4 ml

Physical form

Lyophilized powder stabilized with lactose

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?