• USA Home
  • T5051 - Trimethoxy(vinyl)silane

EMAIL THIS PAGE TO A FRIEND
T5051 Sigma-Aldrich

Trimethoxy(vinyl)silane

≥97%

Synonym: (Trimethoxysilyl)ethylene, Ethenyltrimethoxysilan, Vinyltrimethoxysilane

  • CAS Number 2768-02-7

  • Empirical Formula (Hill Notation) C5H12O3Si

  • Molecular Weight 148.23

  •  Beilstein/REAXYS Number 1099136

  •  EC Number 220-449-8

  •  MDL number MFCD00008605

Purchase

Properties

assay   ≥97%
refractive index   n20/D 1.339 (lit.)
density   0.97 g/mL at 25 °C (lit.)
  0.971 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   CO[Si](OC)(OC)C=C
Safety & Documentation

Safety Information

RIDADR 
UN 1993 3/PG 2
WGK Germany 
1
RTECS 
VV6770000
Flash Point(F) 
72 °F
Flash Point(C) 
22 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

235768 Vinyltrimethoxysilane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?