• USA Home
  • T5692 - Monoclonal Anti-TCF-3/4 antibody produced in mouse

T5692 Sigma

Monoclonal Anti-TCF-3/4 antibody produced in mouse

~0.5 mg/mL, clone 6F12-3, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-T-Cell Factor 3/4



Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Gene Regulation, Antibodies for Hematopoietic Stem Cells,
species reactivity   mouse, human
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  western blot: 1-4 μg/mL using whole extracts of Cos-1 cells transfected with human TCF-3
clone   6F12-3, monoclonal
concentration   ~0.5 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 60 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   mouse ... Tcf3(21415)
biological source   mouse
conjugate   unconjugated



His-tagged fusion protein corresponding to amino acids 31-331 of human TCF.

Physical form

Supplied as a 0.2 μm sterile-filtered solution in 0.1 M Tris-glycine, pH 7.4, 0.15 M sodium chloride, and 30% glycerol with 0.08% sodium azide.

Price and Availability

Antibody Explorer

Explore with Confidence

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, full-face respirator (US), Gloves, multi-purpose combination respirator cartridge (US), type ABEK (EN14387) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?