• USA Home
  • T5762 - 3-(Trimethylsilyl)propionic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
T5762 Sigma

3-(Trimethylsilyl)propionic acid sodium salt

≥99%

Purchase

Properties

assay   ≥99%
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?