• USA Home
  • T6033 - Sodium p-toluenesulfinate

EMAIL THIS PAGE TO A FRIEND
T6033 Sigma-Aldrich

Sodium p-toluenesulfinate

Synonym: p-Toluenesulfinic acid sodium salt

  • CAS Number 824-79-3

  • Linear Formula 4-CH3-C6H4SO2Na

  • Molecular Weight 178.18

  •  Beilstein/REAXYS Number 3574491

  •  EC Number 212-538-5

  •  MDL number MFCD00013136

  •  PubChem Substance ID 24900340

Purchase

Properties

SMILES string   [Na+].Cc1ccc(cc1)S([O-])=O
InChI   1S/C7H8O2S.Na/c1-6-2-4-7(5-3-6)10(8)9;/h2-5H,1H3,(H,8,9);/q;+1/p-1
InChI key   KFZUDNZQQCWGKF-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XT4725000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

455652 Sodium p-toluenesulfinate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?