• USA Home
  • T7428 - Topoisomerase I from wheat germ

EMAIL THIS PAGE TO A FRIEND
T7428 Sigma-Aldrich

Topoisomerase I from wheat germ

buffered aqueous glycerol solution

Synonym: DNA relaxing enzyme, Topo I

Purchase

Properties

Related Categories 5.99.x.x Others, 5.x.x.x Isomerases, Application Index, Biochemicals and Reagents, Cancer Research,
grade   for molecular biology
form   buffered aqueous glycerol solution
mol wt   97 kDa
foreign activity   RNase and DNase, none detected
shipped in   dry ice
storage temp.   −70°C

Description

General description

Topoisomerase I relaxes supercoiled DNA molecules. The enzyme initiates transient breakages and rejoins of phosphodiester bonds in superhelical turns of closedcircular dsDNA.

Application

Suitable for:
• Analsis of degree of DNA supercoiling
• Forming knots and circles in single stranded DNA
• Converting complementary single stranded DNA to double stranded, circular forms
• Rejoining nicks in double-stranded DNA

Biochem/physiol Actions

Topoisomerase I relaxes supercoiled DNA molecules. The enzyme initiates transient breakages and rejoins of phosphodiester bonds in superhelical turns of closed-circular DNA. Enzyme activity is independent of right- and left-handed superhelices.

Components

Topoisomerase I is supplied in a solution of 50 mM Tris-HCl, pH 7.9, 1 mM EDTA, 1 mM DTT, 0.5 M NaCl and 50% glycerol (v/v).

Unit Definition

One unit will convert 1μg of supercoiled pGEM9Zf(−) plasmid DNA to the relaxed form in 30 min at 37 °C.

Other Notes

To preserve enzyme activity, avoid multiple freeze-thaw cycles and frequent temperature variations.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Metabolomics, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

N9653 Netropsin dihydrochloride, from Streptomyces netropsis, ≥98% (HPLC and TLC), powder
L2037 β-Lapachone, ≥98% (TLC)
K0133 Kaempferol, ≥90% (HPLC), powder

related product

Product #

Description

Add to Cart

LIG1 DNA Ligation Kit
D9380 DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
D8276 DNA Polymerase I, Klenow Fragment from Escherichia coli, buffered aqueous glycerol solution
D2886 DNA Ligase from T4-infected Escherichia coli, buffered aqueous glycerol solution
P4978 Phosphatase, Alkaline from calf intestine, buffered aqueous glycerol solution
P4390 Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution
G5663 Glutathione S-Transferase from E. coli, recombinant, expressed in E. coli, buffered aqueous solution
D7291 Deoxyribonuclease I RNase-free solution from bovine pancreas
R6513 Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
R4642 Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?