• USA Home
  • T8312 - Anti-Tissue Inhibitor of Metalloproteinase-4, C-Terminal antibody produced in rabbit

T8312 Sigma

Anti-Tissue Inhibitor of Metalloproteinase-4, C-Terminal antibody produced in rabbit

~1 mg/mL, buffered aqueous glycerol solution, affinity isolated antibody

Synonym: Anti-TIMP-4



Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Extracellular Matrix, Antibodies to Matrix Metalloproteinases and TIMPs,
species reactivity   human
application(s)   western blot: 1:1,000 using a concentrated cell culture medium from stimulated human endothelial cells
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... TIMP4(7079)
biological source   rabbit
conjugate   unconjugated



synthetic peptide corresponding to the C-terminal end of human tissue inhibitor of metalloproteinase-4

Physical form

In 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.


By immunoblotting against the reduced protein, the antibody reacts with bands at approximately 23 kDa and 29 kDa (unglycosylated and glycosylated, respectively). It also reacts with non-reduced TIMP-4.

Price and Availability

Explore with Confidence

Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Safety & Documentation

Safety Information

Symbol  GHS08  GHS08
Signal word  Danger
Hazard statements  H319-H334
Precautionary statements  P261-P305 + P351 + P338-P342 + P311
Personal Protective Equipment  Eyeshields, Faceshields, full-face respirator (US), Gloves, multi-purpose combination respirator cartridge (US), type ABEK (EN14387) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy


Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?