• USA Home
  • T8324 - Triisopropylnaphthalenesulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
T8324 Sigma-Aldrich

Triisopropylnaphthalenesulfonic acid sodium salt

suitable for RNA purification

Synonym: TIPS

  • CAS Number 1323-19-9

  • Empirical Formula (Hill Notation) C19H25NaO3S

  • Molecular Weight 356.45

  •  EC Number 215-351-7

Purchase

Properties

application(s)   RNA purification: suitable

Description

Application

Used in RNA extractions.

Analysis Note

Contains a mixture of isomers and other synthetic by-products.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?