EMAIL THIS PAGE TO A FRIEND
T8777 Sigma-Aldrich

1-Triacontanol

suitable for plant cell culture, ~99% (capillary GC)

Synonym: 1-Hydroxytriacontane, Melissyl alcohol

  • CAS Number 593-50-0

  • Linear Formula CH3(CH2)28CH2OH

  • Molecular Weight 438.81

  •  Beilstein/REAXYS Number 1711965

  •  EC Number 209-794-5

  •  MDL number MFCD00002963

Purchase

Properties

assay   ~99% (capillary GC)
application(s)   cell culture | plant: suitable
mp   86-87 °C (lit.)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO
InChI   1S/C30H62O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31/h31H,2-30H2,1H3
InChI key   REZQBEBOWJAQKS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?