• USA Home
  • T9076 - Monoclonal Anti-TCP-1β antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
T9076 Sigma-Aldrich

Monoclonal Anti-TCP-1β antibody produced in mouse

~2 mg/mL, clone F39P7F11, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-CCT2, Anti-CCTB, Anti-Chaperonin containing T-complex polypeptide 1β subunit, Anti-T-Complex Protein 1β

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to Chaperonins and Heat Shock Proteins,
conjugate   unconjugated
clone   F39P7F11, monoclonal
biological source   mouse
application(s)   immunohistochemistry: suitable
  western blot: 1-2 μg/mL using HeLa total cell extract
species reactivity   rat, mouse, monkey, bovine, canine, human
mol wt   antigen ~50 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG2a
antibody product type   primary antibodies
concentration   ~2 mg/mL
UniProt accession no.   P78371
Gene Information   human ... CCT2(10576)
mouse ... Cct2(12461)
rat ... Cct2(299809)

Description

Immunogen

synthetic peptide corresponding to amino acids 526-535 of human TCP1β. This sequence is identical in bovine, rat, mouse, and frog.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?