EMAIL THIS PAGE TO A FRIEND
T9125 Sigma-Aldrich

Thulium(III) oxide

Purchase

Properties

SMILES string   O=[Tm]O[Tm]=O
InChI   1S/3O.2Tm
InChI key   CWEYWFRFCUNVEY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

289167 Thulium(III) oxide, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?