• USA Home
  • U21108-HH - DMT-2′O-TBDMS-rU Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
U21108-HH Sigma-Aldrich

DMT-2′O-TBDMS-rU Phosphoramidite

configured for PerkinElmer 8900, configured for Polygen

Synonym: 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-uridine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-2′-O-TBDMS-rU amidite

Purchase

Properties

Related Categories 2′O-TBDMS RNA Phosphoramidites, Chemical Synthesis, Expedite, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents), Synthesizer Material Cross Reference More...
biological source   non-animal source (no BSE/TSE risk)
InChI Key   SKNLXHRBXYGJOC-ZMHKPELYSA-N
product line   Proligo Reagents
assay   ≥99% (31P-NMR)
  ≥99.0% (reversed phase HPLC)
  conforms (1H-NMR)
  conforms (LC/MS)
form   powder
packaging   pkg of 1 × 0.5 g (U211081-01)
application(s)   oligo synthesis: suitable
impurities   ≤0.3 wt. % water content (Karl Fischer)
  ≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
  ≤0.5% single Impurity (reversed phase HPLC)
  ≤3 wt. % residual Solvent content
color   white to off-white
λ   conforms (UV/VIS Identity)
compatibility   configured for PerkinElmer 8900
  configured for Polygen
nucleoside profile   base: uridine
base protecting group: none
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast/standard
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?