• USA Home
  • U21123-HH - DMT-2′Fluoro-dU Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
U21123-HH Sigma-Aldrich

DMT-2′Fluoro-dU Phosphoramidite

configured for ABI

Synonym: DMT-2′-Fluoro-dU amidite

Purchase

Properties

Related Categories 2′Fluoro Phosphoramidites, ABI 3900, ABI 39X, Chemical Synthesis, DMT-2′Fluoro-dU Phosphoramidite,
biological source   non-animal source (no BSE/TSE risk)
InChI Key   HQHQPAYRJJMYQX-DJTPZYMWSA-N
product line   Proligo Reagents
assay   ≥99% (31P-NMR)
  ≥99.0% (reversed phase HPLC)
  conforms (1H-NMR)
  conforms (LC/MS)
form   powder
packaging   pkg of 1 × 0.25 g U211231-01
  pkg of 1 × 1 g U211233-01
application(s)   oligo synthesis: suitable
impurities   ≤0.3% water content (Karl Fischer)
  ≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
  ≤0.5% single Impurity (reversed phase HPLC)
  ≤3% residual Solvent content
color   white to off-white
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
nucleoside profile   base: deoxyuridine
base protecting group: none
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?