• USA Home
  • U60108-HH - DMT-rU-CPG column

EMAIL THIS PAGE TO A FRIEND
U60108-HH Sigma-Aldrich

DMT-rU-CPG column

pore size 500 Å, 1 μmol, configured for ABI

Purchase

Properties

Related Categories ABI 39X, Chemical Synthesis, Column for ABI 39X and 3400, Columns for Synthesizer, Controlled Pore Glass (CPG),
product line   Proligo Reagents
assay   ≥98.0% (Nucleoside Purity, reversed phase HPLC)
packaging   pkg of 1 × 4 columns
pore size   500 Å
capacity   ≥95.0 % (Coupling Efficiency)
λ   conforms (UV/VIS Identity)
compatibility   configured for ABI
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?