EMAIL THIS PAGE TO A FRIEND
UC210 Sigma-Aldrich

8-Hydroxywarfarin

Synonym: 3-(α-Acetonylbenzyl)-4,8-dihydrocoumarin, 4-8-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one

  • CAS Number 17834-04-7

  • Empirical Formula (Hill Notation) C19H16O5

  • Molecular Weight 324.33

  •  MDL number MFCD00272667

  •  PubChem Substance ID 329827659

  •  NACRES NA.77

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Drug and Xenobiotic Metabolism, Multi-Drug Resistance and Drug Metabolism, Substrates, Inducers, Inhibitors More...
Quality Level   100
color   white
mp   185-187 °C
storage temp.   2-8°C
SMILES string   CC(=O)CC(c1ccccc1)C2=C(O)c3cccc(O)c3OC2=O
InChI   1S/C19H16O5/c1-11(20)10-14(12-6-3-2-4-7-12)16-17(22)13-8-5-9-15(21)18(13)24-19(16)23/h2-9,14,21-22H,10H2,1H3
InChI key   BHBOXPNWDGYJNB-UHFFFAOYSA-N

Description

Application

8-Hydroxywarfarin has been used for developing assays for chromatographic separation of hydroxylated warfarin isomers.

Biochem/physiol Actions

Minor CYP1A2 metabolite of (R)-warfarin

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?