• USA Home
  • V0265 - Anti-VHL (200-213) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
V0265 Sigma-Aldrich

Anti-VHL (200-213) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-HRCA1, Anti-RCA1, Anti-VHL1, Anti-Von Hippel-Lindau tumor suppressor isoform 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, V
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1:500-1:2,000
species reactivity   human
mol wt   antigen ~24 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
UniProt accession no.   P40337
Gene Information   human ... VHL(7428)

Description

Immunogen

synthetic peptide corresponding to amino acids 200-213 of human VHL

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Oncogenes and Tumor Suppressors Reprogram Metabolism

Proliferating cells require the biosynthesis of structural components for biomass production and for genomic replication. This requires a reprogramming of the metabolic pathways to ensure nutrients s...
BioFiles v7 n4
Keywords: Aerobic, Antitumor agents, Apoptosis, Cancer, Citric Acid Cycle, Degradations, Environmental, Gene expression, Glycolysis, Growth factors, Metabolic Pathways, Metabolism, Metabolites, Nucleotide Synthesis, Pentose phosphate pathway, Phosphorylations

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?