• USA Home
  • V1765 - Anti-VRK1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
V1765 Sigma-Aldrich

Anti-VRK1 antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Vaccinia related kinase 1, Anti-Vaccinia virus B1R-related kinase 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Primary Antibodies,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 2.0-4.0 μg/mL using HEK-293T cell lysate expressing human VRK1
species reactivity   human
mol wt   antigen ~45 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~1.5 mg/mL
UniProt accession no.   Q99986
Gene Information   human ... VRK1(7443)
mouse ... Vrk1(22367)
rat ... Vrk1(362779)

Description

General description

VRK1 is serine/threonine kinase that regulates nuclear condensation, mRNA synthesis and cell cycle. This kinase also functions as an upstream regulator of p53 and phosphorylates p53 in Thr18. Phosphorylation of VRK1 in Ser342 by Plk3 induces the fragmentation of Golgi apparatus . Anti-VRK1 antibody is specific for human VRK1 (approx. 45 kDa). In immunoblotting, detection of the VRK1 band is specifically inhibited by the VRK1 immunizing peptide.

Application

Anti-VRK1 is suitable for use in immunoprecipitation using HEK-293T cell lysates . The product may also be used for western blot (2.0-4.0 μg/mL using HEK-293T cell lysate expressing human VRK1).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?