• USA Home
  • V2251 - Snake venom from Vipera ammodytes (Long-nosed viper)

EMAIL THIS PAGE TO A FRIEND
V2251 Sigma-Aldrich

Snake venom from Vipera ammodytes (Long-nosed viper)

Synonym: Southern European Sand Viper

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?