• USA Home
  • V5253 - Val-Asp-[Arg8]-Vasopressin

EMAIL THIS PAGE TO A FRIEND
V5253 Sigma-Aldrich

Val-Asp-[Arg8]-Vasopressin

≥95% (HPLC)

  • CAS Number 100930-18-5

  • Empirical Formula (Hill Notation) C55H78N16O17S2

  • Molecular Weight 1299.43

  •  MDL number MFCD00076742

Purchase

Properties

assay   ≥95% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C(N)C(=O)NC(CC(O)=O)C(=O)NC1CSSCC(NC(=O)C(CC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C(Cc2ccccc2)NC(=O)C(Cc3ccc(O)cc3)NC1=O)C(=O)N4CCCC4C(=O)NC(CCCNC(N)=N)C(=O)NCC(O)=O

Description

Amino Acid Sequence

Val-Asp-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2 [Disulfide Bridge: 1-6]

Biochem/physiol Actions

Has natriuretic activity equivalent to Arg8-vasopressin with low pressor activity

Other Notes

Originally isolated from bovine posterior pituitary, thought to be a storage form of Arg8-vasopressin

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?