• USA Home
  • WH0000439M3 - Monoclonal Anti-ASNA1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
WH0000439M3 Sigma-Aldrich

Monoclonal Anti-ASNA1 antibody produced in mouse

clone 2H3, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-ARSA1, Anti-ARSAI, Anti-MGC3821, Anti-arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)

  •  NACRES NA.41

Purchase

Properties

Related Categories AS-ATG, Alphabetical Index, Antibodies, Antibodies to Drug Transport Proteins, Biochemicals and Reagents,
conjugate   unconjugated
clone   2H3, monoclonal
biological source   mouse
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  indirect ELISA: suitable
  indirect immunofluorescence: suitable
  western blot: 1-5 μg/mL
species reactivity   mouse, rat, human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1κ
Quality Level   100
antibody product type   primary antibodies
GenBank® accession no.   NM_004317
UniProt accession no.   O43681
Gene Information   human ... ASNA1(439)

Description

General description

ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.

TMD recognition complex of 40 kDa (TRC40) also known as arsA arsenite transporter, ATP-binding, homolog 1 (ASNA1), is encoded by the gene mapped to human chromosome 19p13.2. ASNA1 is the mammalian ortholog of yeast Arr4/Get3 gene. The encoded protein is abundantly expressed in pancreatic β cells.

Immunogen

ASNA1 (NP_004308, 239 a.a. ~ 348 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PEQTTFICVCIAEFLSLYETERLIQELAKCKIDTHNIIVNQLVFPDPEKPCKMCEARHKIQAKYLDQMEDLYEDFHIVKLPLLPHEVRGADKVNTFSALLLEPYKPPSAQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Biochem/physiol Actions

Asna1/TRC40 interacts with the transmembrane domains (TMD) of newly released tail-anchored (TA) proteins and transfers them to the endoplasmic reticulum (ER) receptor for subsequent membrane insertion and thus, plays an essential role in TA protein biogenesis. The encoded protein is necessary for effective HSV1 replication. Asna1 acts as a key regulator of insulin signaling in C. elegans and mammalian cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?