• USA Home
  • WH0004059M1 - Monoclonal Anti-BCAM antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
WH0004059M1 Sigma-Aldrich

Monoclonal Anti-BCAM antibody produced in mouse

clone 2F3, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-AU, Anti-BCAM, Anti-Lutheran blood group (Auberger b antigen included), Anti-MSK19

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, B-BI, Primary Antibodies
conjugate   unconjugated
clone   2F3, monoclonal
biological source   mouse
application(s)   indirect ELISA: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1κ
antibody product type   primary antibodies
immunogen sequence   EVRLSVPPLVEVMRGKSVILDCTPTGTHDHYMLEWFLTDRSGARPRLASAEMQGSELQVTMHDTRGRSPPYQLDSQGRLVLAEAQVGDERDYVCVVRAGA* (without GST)
UniProt accession no.   P50895
Gene Information   human ... BCAM(4059)

Description

Immunogen

LU (NP_005572, a.a. 32-132) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.

Physical form

Solution in phosphate buffered saline, pH 7.2

Safety & Documentation
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?