• USA Home
  • WH0004891M1 - Monoclonal Anti-SLC11A2 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
WH0004891M1 Sigma-Aldrich

Monoclonal Anti-SLC11A2 antibody produced in mouse

clone 4C6, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-DCT1, Anti-DMT1, Anti-NRAMP2, Anti-solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Drug Transport Proteins, Biochemicals and Reagents, Drug Transport Proteins & Reagents,
conjugate   unconjugated
clone   4C6, monoclonal
biological source   mouse
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  indirect ELISA: suitable
  western blot: 1-5 μg/mL
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG2aκ
antibody product type   primary antibodies
Quality Level   100
GenBank® accession no.   NM_000617
UniProt accession no.   P49281
Gene Information   human ... SLC11A2(4891)

Description

General description

The SLC11A2 gene encodes a divalent metal transporter (DMT1), which carries iron, manganese, cobalt, nickel, cadmium, lead, copper, and zinc. DMT1 participates in cellular iron absorption at the luminal surface of the duodenum as well as in other areas of the body (Hubert and Hentze, 2002 [PubMed 12209011]; Ludwiczek et al., 2007 [PubMed 17293870]).[supplied by OMIM

Immunogen

SLC11A2 (NP_000608, 1 a.a. ~ 65 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVLGPEQKMSDDSVSGDHGESASLGNINPAYSNPSLSQSPGDSEEYFATYFNEKISIPEEEYSCF

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?