• USA Home
  • WH0009344M3 - Monoclonal Anti-TAOK2 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
WH0009344M3 Sigma-Aldrich

Monoclonal Anti-TAOK2 antibody produced in mouse

clone 2E2, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-KIAA0881, Anti-MAP3K17, Anti-PSK1, Anti-PSK, Anti-TAO kinase 2, Anti-TAO1, Anti-TAO2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, T-TB
conjugate   unconjugated
clone   2E2, monoclonal
biological source   mouse
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  indirect ELISA: suitable
  western blot: 1-5 μg/mL
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG2aκ
antibody product type   primary antibodies
GenBank® accession no.   BC051798
UniProt accession no.   Q9UL54
Gene Information   human ... TAOK2(9344)

Description

Immunogen

TAOK2 (AAH51798, 831 a.a. ~ 930 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ELQCRQYKRKMLLARHSLDQDLLREDLNKKQTQKDLECALLLRQHEATRELELRQLQAVQRTRAELTRLQHQTELGNQLEYNKRREQELRQKHAAQVRQQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?