• USA Home
  • WH0009775M1 - Monoclonal Anti-DDX48 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
WH0009775M1 Sigma-Aldrich

Monoclonal Anti-DDX48 antibody produced in mouse

clone 2D12, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-DEAD (Asp-Glu-Ala-Asp) box polypeptide 48, Anti-KIAA0111, Anti-MGC10862, Anti-NUK34, Anti-eIF4A3, Anti-eIF4AIII, Anti-hNMP265

Purchase

Properties

clone   2D12, monoclonal
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  indirect ELISA: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1κ
immunogen sequence   PQKERESIMKEFRSGASRVLISTDVWARGLDVPQVSLIINYDLPNNRELYIHRIGRSGRYGRKGVAINFVKNDDIRILRDIEQYYSTQIDEMPMNVADLI* (without GST)
Gene Information   human ... EIF4A3(9775)

Description

Immunogen

DDX48 (NP_055555, a.a. 312-412) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.

Physical form

Solution in phosphate buffered saline, pH 7.2

Safety & Documentation

Safety Information

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?