• USA Home
  • WPB01429WA - Whirl-Pak® High Temperature Bag
EMAIL THIS PAGE TO A FRIEND
WPB01429WA Sigma
Whirl-Pak® High Temperature Bag
532 mL capacity(18 oz)
Synonym: autoclave
Purchase
Properties
Related Categories Bags, Labware More...
sterility   sterile
packaging   pkg of 500 ea
mfr. no.   , B01429WA (Nasco)
W × L × thickness   11.5 cm × 23 cm × 0.076 mm
capacity   532 mL capacity(18 oz)
Description
General description
For hot fill samples with a temperature up to 215° F (102° C), the Whirl-Pak high temperature bag is specially made for this application. The FDA approved film is a co-extrusion of low density polyethylene and nylon that will hold liquids, solids, sauces, gravies, and other samples having a high temperature. The bag is self standing and does not need a rack or special holder to keep it upright, making sample collection safe and easy. The write-on strip provides an easy place to list sample information on the bag. (Do not put bag in boiling water.)
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?