• USA Home
  • WPB01545WA - Whirl-Pak® Detectable Bag
EMAIL THIS PAGE TO A FRIEND
WPB01545WA Sigma
Whirl-Pak® Detectable Bag
532 mL capacity(18 oz)
Purchase
Properties
Related Categories Bags, Labware More...
packaging   pkg of 500 ea
mfr. no.   , B01545WA (Nasco)
W × L × thickness   11.5 cm × 23 cm × 0.064 mm
capacity   532 mL capacity(18 oz)
Description
General description
Steel wire used for bag closure is detectable with standard metal detection equipment. Protect your product and maintain quality with these sampling bags.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?