• USA Home
  • XNAB2E - Extract-N-Amp Blood PCR Kit

EMAIL THIS PAGE TO A FRIEND
XNAB2E Sigma-Aldrich

Extract-N-Amp Blood PCR Kit

sufficient for 100 extractions, sufficient for 500 amplifications

Purchase

Properties

usage   sufficient for 100 extractions
  sufficient for 500 amplifications
shipped in   wet ice
storage temp.   −20°C

Description

General description

The Extract-N-Amp Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by PCR (Fig. 1). This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The kit also includes a PCR ReadyMix, specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start, for specific amplification.

Features and Benefits

• Efficient 8 minute prep allows greater speed and throughput
• No need for any type of purification, organic extraction, centrifugation or alcohol precipitation
• Simple, 3 step procedure with no special equipment required
• Hot start antibody included for highly specific PCR amplification of genomic DNA
• Compatible with multiple format (single tube or 96-well plate)
• Can be used with whole blood or blood cards
• Extract stable at 4 °C for at least 6 months (Fig. 2)

Principle

Genomic DNA is extracted from 10 μl of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR ReadyMix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
UN 3316 9/PG 3
Protocols & Articles

Protocols

Extract-N-Amp™ Blood PCR Kit Protocol

Product Description Reagents and Equipment Required But Not Provided Storage/Stability Precautions and Disclaimer Procedure PCR amplification Troubleshooting Guide Materials References
Keywords: Amplification, Coagulation, Detection methods, Diagnostic, Evaporation, Gas chromatography, Nucleic acid annealing, Nucleic acid denaturation, Polymerase chain reaction, Sequencing, Solvents

Related Content

PCR Selection Guide

We offer a wide variety of PCR enzymes, master mixes, and PCR protocols to meet your experimental needs for routine PCR, qPCR, or RT-PCR. Our PCR Selection Guide features various filters to sort by, ...
Keywords: Genomics, Polymerase chain reaction, Polymerase chain reaction - quantitative

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?