• USA Home
  • Z315664 - Eppendorf® Varispenser® Plus

EMAIL THIS PAGE TO A FRIEND
Z315664

Eppendorf® Varispenser® Plus

volume 10-50 mL, accuracy: 0.5%, resolution: 1.0 mL

Purchase

Properties

Related Categories Dispensers, Eppendorf, Eppendorf Dispensers & Burettes, Eppendorf Liquid Handling, Eppendorf Varispensers & Varispenser Plus,
feature   provided with adapters for 32, 38 and 40 mm threads.
  telescopic intake tube allows use with short or tall bottles.
  fits 45 mm bottle thread
mfr. no.   Eppendorf®, 4961 000.055
volume measuring range   10-50 mL, accuracy: 0.5%, resolution: 1.0 mL
precision   ±0.1%

Description

General description

Bottle-top dispensers with PTFE valve blocks and pistons and a Pt-lr spring bolosilicate ball valve for high resistance to aggressive chemicals. Modular construction for simple cleaning and maintenance.

Safety valve for priming without loss of reagent and fine adjustment of volume.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Varispenser is a registered trademark of Eppendorf AG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?