• USA Home
  • Z324078 - BRAND® Digital Burette III bottle-top burette

EMAIL THIS PAGE TO A FRIEND
Z324078

BRAND® Digital Burette III bottle-top burette

volume 50 mL, accuracy: 0.2%, with SafetyPrime

Purchase

Properties

feature   subdivision 0.01 mL
  with SafetyPrime
Precision measuring range   precision: ±0.1%
volume measuring range   50 mL, accuracy: 0.2%

Description

General description

The Digital Burette III bridges the gap between cumbersome glass burettes and costly titrating units. Its small size and long-life battery allow it to be used in cramped quarters and in locations far from electrical outlets.

The SafetyPrime recirculating valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes.

To allow the bottle label to face the user, the valve block can be rotated 360°. Easy-grip hand wheel action simplifies the continuous regulation of titration rate, from fast to dropwise.

Easy Calibration System for calibration adjustments without tools. Straightforward and intuitive operation together with high reliability simplifies your daily routine, whether in the lab or in the field.

Items supplied:
Digital Burette III, conformity certified with performance certificate, fine tip titrating tube, telescoping filling tube, SafetyPrime recirculation valve (optional), valve mounting tool and PP adapers (GL 45/32, GL 45/S40 & GL 32/NS 29/32).

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

SafetyPrime is a trademark of BRAND GMBH + CO KG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

recommended

Product #

Description

Add to Cart

Z567132 BRAND® Titrette® digital bottle-top burette, volume 50 mL, accuracy: 0.06%, with interface RS 232

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?