• USA Home
  • Z361119 - Slide scanner, SprintScan 35
EMAIL THIS PAGE TO A FRIEND
Z361119 Sigma
Slide scanner, SprintScan 35
for Macintosh
Purchase
Properties
Related Categories Labware, Light Microscopy, Manufacturer Browser, Microscopy & Staining, Polaroid,
AC/DC input   110 / 220, 60-50
mfr. no.   Polaroid
availability   not available in EU
Description
Compatibility
Requires System 7.0.1 or higher; Macintosh II or Quadra with at least 8 MB RAM (16 MB recommended).
Features and Benefits
Create high-resolution digital scans of your 35mm slides in less than one minute. Just set desired resolution from 72 dpi − excellent for screen displays − to 2700 dpi − fine enough for full-page professional printing requirements. Then drop in the slide and point-and-click in the scanning utility software (included). SprintScan uses proprietary CCD technology to capture 24-bit RGB color in a single pass. Scans mounted slides and unmounted film or filmstrips (film holder included). Even negatives can be scanned and "reversed" to display as positive color.
Includes SCSI cable and terminator, and scanning software. Also features Adobe Photoshop plug-in module supporting image-editing and Photoshop PIM architecture. Crop, resize, color-correct, adjust sharpness all with automatic preview update. Film table allows you to store parameters for popular types of film.
Legal Information
SprintScan is a trademark of Polaroid Corp.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?