EMAIL THIS PAGE TO A FRIEND
Z361720 Sigma-Aldrich
TableCurve
3D for Windows 3.1
Purchase
Properties
mfr. no.   AISN
  Spss
availability   not available in EU
Description
General description
Automate the difficult task of finding the best equation to fit experimental data. Import data as ASCII files or from Lotus, Excel, or Quattro Pro spreadsheets. Then TableCurve, in a single processing step, fits the data to all of its built-in equations and ranks them by correlation coefficient. View results as graphs or numeric summaries. Output hardcopy graphs or create a file of fit results. Choose TableCurve 2D if your data involves curve-fitting two-variable (x,y) data; TableCurve 3D for surface-fitting three-variable (x,y,z) data.
TableCurve 3D® gives scientists the power to find the ideal model for even the most complex data, including equations that might never have been considered. TableCurve 3D�s built-in equation set includes a wide array of linear and nonlinear models for any application:
• Linear equations
• Polynominal and rational functions
• Logrithmic and exponential functions
• Nonlinear peak functions
• Nonlinear transition functions
• Nonlinear exponential and power equations
• User-defined functions (up to 15)
TableCurve 3D�s state-of-the-art surface fitting includes capabilities not found in other software packages:
• In addition to standard least squares minimization, TableCurve 3D�s non-linear engine is capable of three different robust estimations: least absolute deviation, Lorentzian minimization and Pearson VII Limit minimization
• Option to change the maximum number of terms permitted when fitting linear equations (minimum 3; maximum 11)
• On systems that support multi-threading, TableCurve 3D�s Background Thread Processing option allows fitting to occur without any form of user input
• Option to set the default term significance anywhere from 1 to 15.
System Requirements
Windows 3.1 with supplied Win32s libraries version 1.30c, Windows 95, 98, 2000, NT 33mhz 486 or higher 8MB RAM (12MB recommended) 5MB free hard disk space
Legal Information
TableCurve is a trademark of AISN Software
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?