• USA Home
  • Z364797 - Millipore® Stericup filtration system
EMAIL THIS PAGE TO A FRIEND
Z364797 Millipore
Millipore® Stericup filtration system
capacity 500 mL, Durapore PVDF membrane, pore size 0.45 μm
Purchase
Properties
material   Durapore PVDF membrane
  polypropylene housing (With polystyrene collar)
  polystyrene housing
feature   cap
  graduations
mfr. no.   Millipore, SCHVU05RE
capacity   500 mL
pore size   0.45 μm
Description
General description
Designed for vacuum filtration of tissue culture media and components, biological fluids, and other aqueous solutions. Polystyrene housing with low protein-binding membrane. Screw cap for sterile storage of filtrate. Packaged 12 per case.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a trademark of Merck KGaA, Darmstadt, Germany
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?