• USA Home
  • Z380504 - CollagenPRO COAT cultureware
EMAIL THIS PAGE TO A FRIEND
Z380504 Sigma-Aldrich
CollagenPRO COAT cultureware
96 well plate
Purchase
Description
Application
A 96-well configuration for the attachment of epithelial, endothelial, muscle, and neuronal cell types. Allows comparative analysis of individual conditions in each well.
General description
Type I collagen is an excellent adhesion factor for many cell types, including epithelial, endothelial, muscle, and neuronal lines. CollagenPRO COAT is highly purified and processed to offer the least immunogenic collagen-coated cultureware currently available. The processing also improves clarity (the coated surfaces are more transparent) and provides a natural environment for attachment and rapid proliferation.
All vessels coated with CollagenPRO COAT have flat bottoms.
Legal Information
CollagenPRO is a trademark of Novagen, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Cell Culture Guide
Milli-Q® Water Purification Solutions
Protocols & Articles
Related Content
Cell Culture Troubleshooting Guide
Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?