• USA Home
  • Z600067 - Hellma® absorption cuvettes, standard cells, Macro
EMAIL THIS PAGE TO A FRIEND
Z600067
Hellma® absorption cuvettes, standard cells, Macro
optical glass, spectral range 320-2,500 nm, pathlength 10 mm, chamber volume 3,500 μL
Purchase
Properties
material   optical glass
description   2 polished windows
mfr. no.   Hellma 100-OS
parameter   320-2,500 nm spectral range
H × W   45 mm × 12.5 mm
chamber volume   3,500 μL
pathlength   10 mm
Description
General description
with PTFE lid
Legal Information
Hellma is a registered trademark of Hellma GmbH
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?