• USA Home
  • Z600792 - Hellma® absorption cuvettes, standard cells, semi Micro
EMAIL THIS PAGE TO A FRIEND
Z600792
Hellma® absorption cuvettes, standard cells, semi Micro
Suprasil® quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 1,400 μL
Purchase
Properties
material   Suprasil® quartz
description   2 polished windows
mfr. no.   Hellma 114-QS
availability   not available in USA and Canada
parameter   200-2,500 nm spectral range
chamber volume   1,400 μL
pathlength   10 mm
Description
General description
with PTFE stopper
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?