• USA Home
  • Z606855 - Eppendorf® electroporation cuvettes
EMAIL THIS PAGE TO A FRIEND
Z606855
Eppendorf® electroporation cuvettes
capacity 400 μL, electrode distance 2 mm
Purchase
Properties
sterility   sterile
mfr. no.   Eppendorf®, 4307 000.593 (European)
  Eppendorf®, 4307 000.593
availability   available only in EU
capacity   400 μL
electrode distance   2 mm
Description
General description
• Plastic cuvette with aluminum electrodes
• Individually wrapped and gamma-radiation sterilized
• Correct positioning in the cuvette holder guaranteed due to design
• Clear gap width identification by lateral cuvette label
• Frosted labeling area for clear experiments
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?