EMAIL THIS PAGE TO A FRIEND
Z662577
BILATEC PCR tubes
volume 0.5 mL, dome cap
Purchase
Properties
material   polypropylene
feature   RNase and DNase free
  autoclavable
  dome cap
packaging   pack of 1000
mfr. no.   313051
availability   available only in EU
volume   0.5 mL
Description
General description
• Extremely thin-walled tubes result in excellent thermo-transfer for better, more reliable PCR.
• RNase- and DNase-free
• Compatible with most standard thermocyclers
Domed caps for efficient heat transfer from cyclers lid.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?