• USA Home
  • Z694525 - Nanofiber multiwell plate inserts, with random oriented nanofibers

EMAIL THIS PAGE TO A FRIEND
Z694525 Sigma

Nanofiber multiwell plate inserts, with random oriented nanofibers

for 24 well plates

Synonym: 3D Cell Culture, 3D, 3d plates, tissue culture plates

Purchase

Properties

Related Categories 3D Cell Culture Products, 3D Scaffolds, Cell Culture Supplies, Labware, Nanofiber Scaffolds More...
material   clear wells flat
  transparent plate
sterility   sterile
feature   lid
mfr. no.   Nanofiber Solutions, 122401
size   24 wells
color   transparent
matrix active group   Non-treated surface treatment
suitability   suitable for (Tissue Engineering, Cancer Research)

Description

General description

Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.

The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.

The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Nanofiber Solutions 3D Cell Culture Technology [VIDEO]

Sigma-Aldrich presents a short video regarding the benefits of the 3-D Cell Culture line from Nanofiber Solutions.
Keywords: Cell culture

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?