• USA Home
  • Z703613 - Membrane Microdomain Signaling: Lipid Rafts in Biology and Medicine
EMAIL THIS PAGE TO A FRIEND
Z703613 Sigma-Aldrich
Membrane Microdomain Signaling: Lipid Rafts in Biology and Medicine
  •  ISBN-10 1-58829354-8
  •  ISBN-13 978-1-58829354-1
Purchase
Properties
Related Categories Books and Software, Cell Signaling Books, Labware More...
publication info   M. Mattson, Humana Press, 2004, 224 pp., hard cover
mfr. no.   Humana
Description
General description
Cutting-edge articles review the current understanding of lipid microdomain signaling mechanisms and their physiological and pathological importance. The book describes the role of lipid rafts in learning, memory, and cancer, presents the emerging evidence that lipid rafts play critical roles in signaling pathways and the regulation of synaptic function in the nervous system, and shows how alterations in lipid raft metabolism are implicated in the pathogenesis of neurodegenerative disorders. Techniques are also described for the isolation of lipid rafts, the analysis of the lipid and protein components of lipid rafts, the imaging of lipid rafts in living cells, and the analysis of signal transduction in lipid rafts.
Table of Contents
1. From Intramolecular Asymmetries to Raft Assemblies
2. Regulatory Aspects of Membrane Microdomain (Raft) Dynamics in Live Cells: a Biophysical Approach
3. Lipid Raft Membrane Skeletons
4. Role of Cholesterol in Membrane Microdomain Signaling
5. Raft Lipid Metabolism in Relation to Alkyl-Lysophospholipid Induced Apoptosis
6. Role of Lipid Rafts in Signal Transduction and Synaptic Plasticity of Neural Cells
7. Role of Rafts in Virus Fusion and Budding
8. Alterations in Raft Lipid Meatbolism in Aging and Neurodegenerative Disorders
9. Caveolin and Cancer: A Complex Relationship
10. Dietary Modulation of Lipid Rafts
Index
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?